Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DL5/CD158f Antibody (UP-R1), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB002791 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002791 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002791 Supplier Novus Biologicals Supplier No. NBP262222UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIR2DL5/CD158f Monoclonal specifically detects KIR2DL5/CD158f in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen KIR2DL5/CD158f
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone UP-R1
Conjugate DyLight 350
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD158F, CD158F1, killer cell immunoglobulin-like receptor 2DL5A, killer cell immunoglobulin-like receptor KIR2DL5A, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A, killer-cell Ig-like receptor, KIR2DL5, KIR2DL5.1, KIR2DL5.3, KIR2DL5B
Gene Symbols KIR2DL5A
Host Species Mouse
Immunogen Human CD158f-Ig fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 57292
Test Specificity The antibody MEM-59 recognizes neuraminidase-sensitive epitope on CD43 (Leukosialin), a 95-135 kDa type I transmembrane glycoprotein (mucin-type) which is involved in lymphocyte activation. CD43 is expressed by platelets and at high levels on the surface of all leukocytes; it is negative on resting B lymphocytes and erythrocytes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.