Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DL5/CD158f Antibody (UP-R1), HRP, Novus Biologicals™
SDP

Catalog No. NB343070 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343070 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343070 Supplier Novus Biologicals Supplier No. NBP262222H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIR2DL5/CD158f Monoclonal antibody specifically detects KIR2DL5/CD158f in Human samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen KIR2DL5/CD158f
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone UP-R1
Conjugate HRP
Dilution Flow Cytometry, Immunocytochemistry
Formulation PBS
Gene Alias CD158F, CD158F1, killer cell immunoglobulin-like receptor 2DL5A, killer cell immunoglobulin-like receptor KIR2DL5A, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A, killer-cell Ig-like receptor, KIR2DL5, KIR2DL5.1, KIR2DL5.3, KIR2DL5B
Host Species Mouse
Immunogen Human CD158f-Ig fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 57292
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.