Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DS1/CD158h Antibody (1127B), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB345958 Shop All R&D Systems Products
missing translation for 'changeView'
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345958 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. NB345958 Supplier Novus Biologicals missing translation for 'supplierNo' FAB8887MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

KIR2DS1/CD158h Monoclonal specifically detects KIR2DS1/CD158h in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR2DS1/CD158h
Applications Flow Cytometry
Classification Monoclonal
Clone 1127B
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias killer cell immunoglobulin-like receptor 2DS1, CD158a, CD158H, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 1, p50.1
Host Species Rabbit
Immunogen HEK293 human embryonic kidney cell line transfected with human KIR2DS1/CD158h, Met1-His245, Accession # Q14954
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3806
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.