Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DS4/CD158i Antibody (179315), HRP, Novus Biologicals™
SDP

Catalog No. NB175412 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175412 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175412 Supplier Novus Biologicals Supplier No. FAB1847H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIR2DS4/CD158i Monoclonal specifically detects KIR2DS4/CD158i in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen KIR2DS4/CD158i
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 179315
Conjugate HRP
Formulation PBS
Gene Alias CD158 antigen-like family member I, CD158I, CD158i antigen, cl-39, killer cell immunoglobulin-like receptor 2DS4, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail4,nkat8, killer inhibitory receptor 4-1-2, KIR antigen 2DS4, KKA3KIR1D, MGC120019, MGC125315, MGC125317, MHC class I NK cell receptor, natural killer cell inhibitory receptor, Natural killer-associated transcript 8, NKAT-8, NKAT8KIR412, P58 natural killer cell receptor clones CL-39/CL-17, p58 NK receptor CL-39/CL-17
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR2DS4, Accession # P43632
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3809
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.