Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DS4/CD158i Antibody (5F2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP104314C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP104314C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP104314C Supplier Novus Biologicals Supplier No. NBP104314C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIR2DS4/CD158i Monoclonal specifically detects KIR2DS4/CD158i in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen KIR2DS4/CD158i
Applications Western Blot, ELISA
Classification Monoclonal
Clone 5F2
Conjugate DyLight 650
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CD158 antigen-like family member I, CD158I, CD158i antigen, cl-39, killer cell immunoglobulin-like receptor 2DS4, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail4,nkat8, killer inhibitory receptor 4-1-2, KIR antigen 2DS4, KKA3KIR1D, MGC120019, MGC125315, MGC125317, MHC class I NK cell receptor, natural killer cell inhibitory receptor, Natural killer-associated transcript 8, NKAT-8, NKAT8KIR412, P58 natural killer cell receptor clones CL-39/CL-17, p58 NK receptor CL-39/CL-17
Gene Symbols KIR2DS4
Host Species Mouse
Immunogen Recombinant human KIR2DS4/CD158i (23-223aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3809
Test Specificity Anti-human KIR2DS4 mAb, clone 5F2, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human KIR2DS4 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.