Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DS5/CD158g Antibody (1165A), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB345926 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345926 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345926 Supplier Novus Biologicals Supplier No. FAB8920MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

KIR2DS5/CD158g Monoclonal specifically detects KIR2DS5/CD158g in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR2DS5/CD158g
Applications Flow Cytometry
Classification Monoclonal
Clone 1165A
Conjugate mFluor Violet 450 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias killer cell immunoglobulin-like receptor 2DS5, CD158G, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5, NKAT9
Host Species Rabbit
Immunogen NS0 mouse myeloma cell line transfected with human KIR2DS5/CD158g, His22-His245, Accession # Q14953
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3810
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.