Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DS5/CD158g Antibody (1165A), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB310626 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310626 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310626 Supplier Novus Biologicals Supplier No. FAB8920PEATT594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

KIR2DS5/CD158g Monoclonal antibody specifically detects KIR2DS5/CD158g in Human samples. It is validated for Flow Cytometry

Specifications

Antigen KIR2DS5/CD158g
Applications Flow Cytometry
Classification Monoclonal
Clone 1165A
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias killer cell immunoglobulin-like receptor 2DS5, CD158G, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5, NKAT9
Host Species Rabbit
Immunogen NS0 mouse myeloma cell line transfected with human KIR2DS5/CD158g, His22-His245, Accession # Q14953
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3810
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.