Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir3.1 Antibody (OTI1G3), PerCP, Novus Biologicals™
SDP

Catalog No. NB271065PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271065PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271065PCP Supplier Novus Biologicals Supplier No. NBP271065PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kir3.1 Monoclonal specifically detects Kir3.1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Kir3.1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1G3
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias G protein-activated inward rectifier potassium channel 1, GIRK-1, GIRK1Kir3.1, Inward rectifier K(+) channel Kir3.1, inward rectifier K+ channel KIR3.1, KGA, KIR3.1, Potassium channel, inwardly rectifying subfamily J member 3, potassium inwardly-rectifying channel, subfamily J, member 3
Gene Symbols KCNJ3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 279-501 of human Kir3.1 (NP_002230) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3760.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.