Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR3DL2/CD158k Antibody (539304), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB363895 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363895 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363895 Supplier Novus Biologicals Supplier No. FAB2878K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIR3DL2/CD158k Monoclonal specifically detects KIR3DL2/CD158k in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen KIR3DL2/CD158k
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 539304
Conjugate DyLight 488
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias killer cell immunoglobulin-like receptor 3DL2, CD158K, killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 2, NKAT4, NKAT-4, NKAT4B, p140
Host Species Mouse
Immunogen BaF3 mouse pro-B cell line transfected with human KIR3DL2/CD158k, Accession # P43630
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3812
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.