Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR3DL3/CD158z Antibody (1136B), HRP, Novus Biologicals™
SDP

Catalog No. NB345931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345931 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345931 Supplier Novus Biologicals Supplier No. FAB8919H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

KIR3DL3/CD158z Monoclonal specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen KIR3DL3/CD158z
Applications Flow Cytometry
Classification Monoclonal
Clone 1136B
Conjugate HRP
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD158Z;Killer cell immunoglobulin-like receptor 3DL3;KIR2DL5B;KIR3DL7;KIR44;KIRC1
Host Species Rabbit
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human KIR3DL3/CD158z, Met1-Leu322, Accession # Q8N743
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115653
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.