Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR3DL3/CD158z Antibody (CH21) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB258465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258465 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB258465 Supplier Novus Biologicals Supplier No. NBP321499
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIR3DL3/CD158z Monoclonal antibody specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry, Immunoprecipitation

Specifications

Antigen KIR3DL3/CD158z
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone CH21
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 3-12 ug/ml, Immunoprecipitation
Formulation PBS, pH 7.4
Gene Alias CD158Z;Killer cell immunoglobulin-like receptor 3DL3;KIR2DL5B;KIR3DL7;KIR44;KIRC1
Host Species Mouse
Immunogen Human KIR3DL3/CD158z transfectants
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115653
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.