Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir6.1 Antibody (S366-60), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB259324405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259324405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259324405 Supplier Novus Biologicals Supplier No. NBP259324AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kir6.1 Monoclonal specifically detects Kir6.1 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Kir6.1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S366-60
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ATP-sensitive inward rectifier potassium channel 8, Inward rectifier K(+) channel Kir6.1, inwardly rectifying potassium channel KIR6.1, KIR6.1, Potassium channel, inwardly rectifying subfamily J member 8, potassium inwardly-rectifying channel, subfamily J, member 8, uKATP-1
Gene Symbols KCNJ8
Host Species Mouse
Immunogen Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 3764
Test Specificity Detects 45kDa.
Target Species Human, Mouse, Rat, Gerbil
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.