Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir6.1 Antibody (S366-60), Novus Biologicals™
SDP

Catalog No. NBP259324 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259324 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259324 Supplier Novus Biologicals Supplier No. NBP259324
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

Kir6.1 Monoclonal specifically detects Kir6.1 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Kir6.1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S366-60
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS pH7.4, 50% glycerol with 0.1% Sodium Azide
Gene Alias ATP-sensitive inward rectifier potassium channel 8, Inward rectifier K(+) channel Kir6.1, inwardly rectifying potassium channel KIR6.1, KIR6.1, Potassium channel, inwardly rectifying subfamily J member 8, potassium inwardly-rectifying channel, subfamily J, member 8, uKATP-1
Gene Symbols KCNJ8
Host Species Mouse
Immunogen Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3764
Test Specificity Detects 45kDa.
Target Species Human, Mouse, Rat, Gerbil
Content And Storage Store at -20C.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.