Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir6.2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP100206 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP100206 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP100206 Supplier Novus Biologicals Supplier No. NBP100206
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Kir6.2 Polyclonal specifically detects Kir6.2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Kir6.2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.1 - 0.3 ug/ml, Immunohistochemistry 3 - 5 ug/ml, Immunohistochemistry-Paraffin 3 - 5 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000516.3
Gene Alias ATP-sensitive inward rectifier potassium channel 11, beta-cell inward rectifier subunit, BIRKIR6.2, HHF2, IKATP, Inward rectifier K(+) channel Kir6.2, inwardly rectifying potassium channel KIR6.2, Kir6.2, MGC133230, PHHI, potassium channel inwardly rectifing subfamily J member 11, Potassium channel, inwardly rectifying subfamily J member 11, potassium inwardly-rectifying channel, subfamily J, member 11, TNDM3
Gene Symbols KCNJ11
Host Species Goat
Immunogen Peptide with sequence C-AEDPAKPRYRARQ corresponding to internal region (near N-Terminus) according to NP_000516.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3767
Test Specificity This antibody is expected to recognize isoform 1 (NP_000516.3) only.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.