Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kirrel3/NEPH2 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB081267 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081267 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081267 Supplier Novus Biologicals Supplier No. NBP298153B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kirrel3/NEPH2 Polyclonal specifically detects Kirrel3/NEPH2 in Rat samples. It is validated for ELISA.

Specifications

Antigen Kirrel3/NEPH2
Applications ELISA
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias KIAA1867MGC126824, kin of IRRE like 3 (Drosophila), Kin of irregular chiasm-like protein 3, kin of IRRE-like protein 3, KIRREPRO4502, MRD4, NEPH2MGC126850, nephrin-like 2, Nephrin-like protein 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Kirrel3/NEPH2 (Q09GS6; Met1-Ala523).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84623
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.