Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kirrel3/NEPH2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017311 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017311 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB017311 Supplier Novus Biologicals Supplier No. NBP282000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kirrel3/NEPH2 Polyclonal specifically detects Kirrel3/NEPH2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Kirrel3/NEPH2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 5 ug/mL, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias KIAA1867MGC126824, kin of IRRE like 3 (Drosophila), Kin of irregular chiasm-like protein 3, kin of IRRE-like protein 3, KIRREPRO4502, MRD4, NEPH2MGC126850, nephrin-like 2, Nephrin-like protein 2
Gene Symbols KIRREL3
Host Species Rabbit
Immunogen Kirrel3 antibody was raised against a 19 amino acid peptide near the amino terminus of human Kirrel3. The immunogen is located within amino acids 20 - 70 of Kirrel3. Amino Acid Squence: MAKDKFRRMNEGQVYSFSQ
Molecular Weight of Antigen 60 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84623
Test Specificity Kirrel3/NEPH2 antibody is human, mouse and rat reactive. At least two isoforms are known to exist; this antibody will detect both isoforms. Kirrel3/NEPH2 antibody is predicted to not cross-react with other members of the KIRREL protein family.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.