Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KISS1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP145672 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145672 0.1 mg
NB018573 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP145672 Supplier Novus Biologicals Supplier No. NBP145672

Rabbit Polyclonal Antibody has been used in 5 publications

KISS1 Polyclonal specifically detects KISS1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen KISS1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5 - 4 ug/mL
Formulation PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.03% Proclin 300
Gene Accession No. Q6Y4S4
Gene Alias KiSS-1, KiSS-1 metastasis-suppressor, kisspeptin, Kisspeptin-1, malignant melanoma metastasis-suppressor, metastasis-suppressor KiSS-1, METASTIN, MGC39258
Gene Symbols KISS1
Host Species Rabbit
Immunogen Synthetic peptide surrounding amino acid 17 of mouse KiSS-1 (NP_839991).
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3814
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.