Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KISS1 Mouse anti-Human,Mouse, Unconjugated, Clone: 6A4.27, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | KISS1 |
---|---|
Clone | 6A4.27 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC17512MI
|
MilliporeSigma
MABC175125UL |
25 μL |
Each for $189.29
|
|
|||||
MABC17511MI
|
MilliporeSigma
MABC1751100UL |
100 μL |
Each for $477.80
|
|
|||||
Description
KISS1 Monoclonal antibody specifically detects KISS1 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.Specifications
KISS1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Metastasis-suppressor KiSS-1;Kisspeptin-1 | |
A linear peptide corresponding to 54 amino acids from the C-terminal half of human Kisspeptin-1 (KISS1). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
14.70 kDa calculated. |
6A4.27 | |
Monoclonal | |
Purified | |
Apoptosis and Cancer | |
NP_002247 | |
KISS1, PP5098 | |
IgG1 κ | |
Protein G Purified | |
Clone 6A4.27 is a mouse monoclonal antibody that detects Kisspeptin-1. It targets an epitope with in the KP54 (G68-F121) region. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title