Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIST Antibody (OTI2H4), HRP, Novus Biologicals™
SDP

Catalog No. NBP272383H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272383H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272383H Supplier Novus Biologicals Supplier No. NBP272383H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KIST Monoclonal specifically detects KIST in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen KIST
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2H4
Conjugate HRP
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Gene Alias DKFZp434C1613, EC 2.7.11.1, FLJ23015, kinase interacting with leukemia-associated gene (stathmin), Kinase interacting with stathmin, KISKIS protein kinase, KIST, serine/threonine-protein kinase Kist, U2AF homology motif (UHM) kinase 1, U2AF homology motif kinase 1
Gene Symbols UHMK1
Host Species Mouse
Immunogen Full length recombinant protein of human KIST (NP_787062) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 127933.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.