Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIZUNA Antibody, Novus Biologicals™
SDP

Catalog No. NB10040837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10040837 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10040837 Supplier Novus Biologicals Supplier No. NB10040837

Rabbit Polyclonal Antibody

KIZUNA Polyclonal specifically detects KIZUNA in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen KIZUNA
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_060944.2
Gene Alias C20orf19, centrosomal protein kizuna, chromosome 20 open reading frame 19, DKFZp586H021, HT013, KIZ, Kizuna, MGC102941, MGC141930, NCRNA00153, polo-like kinase 1 substrate 1non-protein coding RNA 153
Gene Symbols PLK1S1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 275 and 325 of human KIZUNA (C20orf19) using the numbering given in entry NP_060944.2 (GeneID 55857).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55857
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.