Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIZUNA Antibody, Novus Biologicals™
SDP

Catalog No. NBP217065 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP217065 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP217065 Supplier Novus Biologicals Supplier No. NBP217065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIZUNA Polyclonal specifically detects KIZUNA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen KIZUNA
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.46mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q2M2Z5
Gene Alias C20orf19, centrosomal protein kizuna, chromosome 20 open reading frame 19, DKFZp586H021, HT013, KIZ, Kizuna, MGC102941, MGC141930, NCRNA00153, polo-like kinase 1 substrate 1non-protein coding RNA 153
Gene Symbols PLK1S1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KIZUNA. The exact sequence is proprietary.
Molecular Weight of Antigen 75 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55857
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.