Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klebsiella Aerogenes Antibody (73/28), Novus Biologicals™
SDP

Catalog No. NB10064558 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064558 0.1 mg
NB017607 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064558 Supplier Novus Biologicals Supplier No. NB10064558
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Klebsiella Aerogenes Monoclonal specifically detects Klebsiella Aerogenes in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Klebsiella Aerogenes
Applications ELISA
Classification Monoclonal
Clone 73/28
Concentration 2.3 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Alias k aerogenes, K. aerogenes
Host Species Mouse
Immunogen Native Klebsiella Aerogenes
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity In a simple ELISA using Biogenesis test groups (BTGs) this is reactive with many serotypes of Klebsiella. Immunohistology: Has detected Klebsiella oxytoca and other serotypes in fresh isolates, including those fixed by phenol, formalin or boiling.
Target Species Bacteria
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.