Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Klebsiella Aerogenes Antibody (73/28), Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Klebsiella Aerogenes |
---|---|
Clone | 73/28 |
Concentration | 2.3 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
Applications | Immunofluorescence, ELISA, Immunocytochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017607
![]() |
Novus Biologicals
NB100645580.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064558
![]() |
Novus Biologicals
NB10064558 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Klebsiella Aerogenes Monoclonal specifically detects Klebsiella Aerogenes in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Klebsiella Aerogenes | |
2.3 mg/mL | |
Immunofluorescence, ELISA, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Bacteria | |
IgG2a | |
Protein A purified | |
In a simple ELISA using Biogenesis test groups (BTGs) this is reactive with many serotypes of Klebsiella. Immunohistology: Has detected Klebsiella oxytoca and other serotypes in fresh isolates, including those fixed by phenol, formalin or boiling. |
73/28 | |
ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Native Klebsiella Aerogenes | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title