Learn More
Kimberly-Clark Professional™ KleenGuard™ A40 Reflex™ Coveralls
Catalog No. 19005124 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Large
2X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-124 -
19-005-126 X-Large
19-005-127 2X-Large
19-005-128 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-005-124 Supplier Kimberly-Clark Professional™ Supplier No. 47995
This item is not returnable. View return policy
KleenGuard™ A40 Reflex Coveralls include a respirator fit hood, zipper front, and elastic wrists and ankles, and feature Reflex sizing for a more comfortable fit and fewer rip outs. They’re made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.
KleenGuard™ A40 Reflex Coveralls™ with Hood comes with a respirator fit hood, zipper front, and elastic wrists and ankles. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates.
These coveralls with hood pass NFPA 99 for antistatic materials, pass ASTM F1670 testing for penetration of blood and bodily fluids, and have Reflex sizing for a more comfortable fit and less rip outs. Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing and so much more.
  • 25 Garments/Case
  • KleenGuard™ A40 Reflex™ Coveralls are made of microporous film laminate with a respirator fit hood, zipper front and elastic wrists and ankles
  • These hooded coveralls offer better liquid and particulate barrier than the competition
  • Made to keep out debris, dry particulates and liquid splashes
  • A40 KleenGuard™ protective suits pass NFPA 99 criteria for antistatic materials