Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF12 Antibody (1230C), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB295565 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295565 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295565 Supplier Novus Biologicals Supplier No. FAB9097T

Rabbit Monoclonal Antibody

KLF12 Monoclonal antibody specifically detects KLF12 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen KLF12
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1230C
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived human KLF12, Asn2-Val402, Accession # Q9Y4X4
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 11278
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.