Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF13 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$170.97 - $418.55
Specifications
| Antigen | KLF13 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 to 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
KLF13 Polyclonal specifically detects KLF13 in Mouse samples. It is validated for Western Blot.Specifications
| KLF13 | |
| Western Blot 1:500 to 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Mouse | |
| Q9Y2Y9 | |
| 51621 | |
| Peptide corresponding to amino acids range from 27-56 of the Kruppel-like factor 13; transcription factor NSLP1; novel Sp1 like zinc finger transcription factor; RANTES factor of late activated T lymphocytes-1 [Homo sapiens]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| basic transcription element binding protein 3, Basic transcription element-binding protein 3, BTEB3FKLF2, BTE-binding protein 3, Krueppel-like factor 13, Kruppel-like factor 13, Novel Sp1-like zinc finger transcription factor 1, NSLP1RFLAT1, RANTES factor of late activated T lymphocytes-1, RANTES factor of late activated T-lymphocytes 1, RFLAT-1FKLF-2, Sp1 like zinc finger transcription factor, Transcription factor BTEB3, Transcription factor NSLP1 | |
| KLF13 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title