Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF13 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP223614 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP223614 0.1 mg
NB019357 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP223614 Supplier Novus Biologicals Supplier No. NBP223614
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLF13 Polyclonal specifically detects KLF13 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen KLF13
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 to 1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q9Y2Y9
Gene Alias basic transcription element binding protein 3, Basic transcription element-binding protein 3, BTEB3FKLF2, BTE-binding protein 3, Krueppel-like factor 13, Kruppel-like factor 13, Novel Sp1-like zinc finger transcription factor 1, NSLP1RFLAT1, RANTES factor of late activated T lymphocytes-1, RANTES factor of late activated T-lymphocytes 1, RFLAT-1FKLF-2, Sp1 like zinc finger transcription factor, Transcription factor BTEB3, Transcription factor NSLP1
Gene Symbols KLF13
Host Species Rabbit
Immunogen Peptide corresponding to amino acids range from 27-56 of the Kruppel-like factor 13; transcription factor NSLP1; novel Sp1 like zinc finger transcription factor; RANTES factor of late activated T lymphocytes-1 [Homo sapiens].
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51621
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.