Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF13 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB318910 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318910 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318910 Supplier Novus Biologicals Supplier No. NBP298809MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLF13 Polyclonal antibody specifically detects KLF13 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen KLF13
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias basic transcription element binding protein 3, Basic transcription element-binding protein 3, BTEB3FKLF2, BTE-binding protein 3, Krueppel-like factor 13, Kruppel-like factor 13, Novel Sp1-like zinc finger transcription factor 1, NSLP1RFLAT1, RANTES factor of late activated T lymphocytes-1, RANTES factor of late activated T-lymphocytes 1, RFLAT-1FKLF-2, Sp1 like zinc finger transcription factor, Transcription factor BTEB3, Transcription factor NSLP1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human KLF13.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51621
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.