Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF13 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | KLF13 |
---|---|
Concentration | 0.31 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2131
![]() ![]() |
Proteintech Group Inc
183521AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Anti-RANTES factor of Late Activated T Lymphocytes-1 (RFLAT-1) polyclonal antiserum. RFLAT-1 is a novel protein belonging to the TFIIIA-like zinc finger protein superfamily and binds to the A site of the RANTES (Regulated upon Activation, Normal T cell Expressed and Secreted) chemokine. RANTES is involved in the generation of inflammatory infiltrate and HIV entry into immune cells.Specifications
KLF13 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
Q9Y2Y9 | |
499171, 51621 | |
KLF13 Fusion Protein Ag13200 | |
Primary | |
-20°C | |
KLF13 |
0.31 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BTE binding protein 3, BTEB3, FKLF2, KLF13, Krueppel like factor 13, Kruppel like factor 13, NSLP1, RFLAT 1, RFLAT1, Transcription factor BTEB3, Transcription factor NSLP1 | |
KLF13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title