Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF14 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151804 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151804 25 μg
NB151803 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151804 Supplier Novus Biologicals Supplier No. NBP31192525UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLF14 Polyclonal antibody specifically detects KLF14 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen KLF14
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, Immunohistochemistry 2 - 4 ug/ml
Formulation PBS
Gene Alias basic transcription element-binding protein 5, Kruppel-like factor 14
Host Species Rabbit
Immunogen Synthetic peptide derived from the human KLF14 protein (Uniprot: Q8TD94)
Purification Method Antigen Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136259
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.