Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ KLF15 Antibody Blocking Peptide
Shop All Novus Biologicals Products

Click to view available options
Quantity:
0.1 mg
Specifications
Specifications
Gene ID (Entrez) | 28999 |
Concentration | 1 mg/mL |
Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-24635. For further blocking peptide related protocol, click here . |
Gene Alias | DKFZp779M1320, kidney-enriched Kruppel-like factor, KKLFKidney-enriched krueppel-like factor, Krueppel-like factor 15, Kruppel-like factor 15 |
Gene Symbol | KLF15 |
Quantity | 0.1 mg |
Research Category | Cardiovascular Biology |
Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction