Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF15 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224635SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224635SS 0.025 mg
NBP224635 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224635SS Supplier Novus Biologicals Supplier No. NBP224635SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 7 publications

KLF15 Polyclonal specifically detects KLF15 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen KLF15
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0-2.0 ug/ml, Immunohistochemistry 2-5 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 2-5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9UIH9
Gene Alias DKFZp779M1320, kidney-enriched Kruppel-like factor, KKLFKidney-enriched krueppel-like factor, Krueppel-like factor 15, Kruppel-like factor 15
Gene Symbols KLF15
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of the human KLF15 protein (between amino acids 100-200) [UniProt Q9UIH9]
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28999
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, 95% homologous in mouse and bovine and 90% homologous in rat and pig.
Target Species Human, Mouse, Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.