Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
$207.00 - $426.00
Specifications
| Antigen | KLF15 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1.0-2.0 ug/ml, Immunohistochemistry 2-5 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 2-5 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224635SS
![]() |
Novus Biologicals
NBP224635SS |
0.025 mg |
Each for $207.00
|
|
|||||
NBP224635
![]() |
Novus Biologicals
NBP224635 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
KLF15 Polyclonal specifically detects KLF15 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| KLF15 | |
| Western Blot 1.0-2.0 ug/ml, Immunohistochemistry 2-5 ug/ml, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunohistochemistry-Paraffin 2-5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| DKFZp779M1320, kidney-enriched Kruppel-like factor, KKLFKidney-enriched krueppel-like factor, Krueppel-like factor 15, Kruppel-like factor 15 | |
| KLF15 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, 95% homologous in mouse and bovine and 90% homologous in rat and pig. |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Bovine | |
| Q9UIH9 | |
| 28999 | |
| Synthetic peptide made to an internal portion of the human KLF15 protein (between amino acids 100-200) [UniProt Q9UIH9] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title