Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF17 Antibody (PCRP-KLF17-1G2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB177971 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177971 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177971 Supplier Novus Biologicals Supplier No. NBP314000IR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

KLF17 Monoclonal antibody specifically detects KLF17 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence

Specifications

Antigen KLF17
Applications Western Blot, Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone PCRP-KLF17-1G2
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias FLJ40160, Kruppel-like factor 17, Zfp393, Zinc finger protein 393Krueppel-like factor 17, ZNF393novel zinc-finger protein
Host Species Mouse
Immunogen Recombinant full-length human KLF17protein (Uniprot: Q5JT82)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128209
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.