Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF4 Antibody (4E6), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP121047A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP121047A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP121047A32 Supplier Novus Biologicals Supplier No. NBP121047AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KLF4 Monoclonal specifically detects KLF4 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen KLF4
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 4000000
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut)
Gene Symbols KLF4
Host Species Mouse
Immunogen Recombinant human KLF4 (1-170aa) purified from E. coli (NP_004226.3).
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9314.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.