Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224749 0.1 mg
NBP224749SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224749 Supplier Novus Biologicals Supplier No. NBP224749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 18 publications

KLF4 Polyclonal specifically detects KLF4 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).

Specifications

Antigen KLF4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry reported in scientific literature (PMID 34428252; 30901290), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 34428252), Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.1 ml PBS with 0.05% Sodium Azide
Gene Accession No. O43474
Gene Alias Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut)
Gene Symbols KLF4
Host Species Rabbit
Immunogen A portion of amino acids 1-50 of human KLF4 was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9314
Test Specificity This antibody will detect a number of isoforms of the FUZ protein ranging from 269 amino acids to 418 amino acids. Multiple bands may be seen in Western blotting.
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.