Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 18 publications
$192.94 - $529.95
Specifications
| Antigen | KLF4 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry reported in scientific literature (PMID 34428252; 30901290), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 34428252), Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224749SS
![]() |
Novus Biologicals
NBP224749SS |
0.025 mg |
Each for $192.94
|
|
|||||
NBP224749
![]() |
Novus Biologicals
NBP224749 |
0.1 mg |
Each for $529.95
|
|
|||||
Description
KLF4 Polyclonal specifically detects KLF4 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
| KLF4 | |
| Western Blot 1-3 ug/ml, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry reported in scientific literature (PMID 34428252; 30901290), Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 34428252), Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Primate | |
| O43474 | |
| 9314 | |
| A portion of amino acids 1-50 of human KLF4 was used as the immunogen for this antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| 0.1 ml PBS with 0.05% Sodium Azide | |
| Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut) | |
| KLF4 | |
| IgG | |
| Protein G purified | |
| This antibody will detect a number of isoforms of the FUZ protein ranging from 269 amino acids to 418 amino acids. Multiple bands may be seen in Western blotting. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title