Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF8 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001050 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001050 Supplier Novus Biologicals Supplier No. NB1001050
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

KLF8 Polyclonal specifically detects KLF8 in Human samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen KLF8
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_009181
Gene Alias Basic krueppel-like factor 3, basic kruppel-like factor 3, BKLF3DKFZp686O08126, DXS741, Krueppel-like factor 8, Kruppel-like factor 8, Zinc finger protein 741, ZNF741MGC138314
Gene Symbols KLF8
Host Species Goat
Immunogen Peptide with sequence VDMDKLINNLEVQ-C corresponding to N-Terminus according to NP_009181.2, NP_001152768.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11279
Test Specificity This antibody is expected to recognize isoform 1 (NP_009181.2) and isoform 2 (NP_001152768.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.