Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227418SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227418SS 0.025 mg
NBP227418 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227418SS Supplier Novus Biologicals Supplier No. NBP227418SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLF8 Polyclonal specifically detects KLF8 in Human, Primate samples. It is validated for Western Blot.

Specifications

Antigen KLF8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. O95600
Gene Alias Basic krueppel-like factor 3, basic kruppel-like factor 3, BKLF3DKFZp686O08126, DXS741, Krueppel-like factor 8, Kruppel-like factor 8, Zinc finger protein 741, ZNF741MGC138314
Gene Symbols KLF8
Host Species Rabbit
Immunogen A portion of amino acids 1-50 from human kLF8 was used as the immunogen for this antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11279
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, sumatran orangutan, and white-cheeked gibbon, 89% in rhesus monkey, and 78% in new world monkey.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.