Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLH Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB090274 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB090274 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB090274 Supplier Novus Biologicals Supplier No. NBP299936DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLH Polyclonal specifically detects KLH in Non-species specific samples. It is validated for ELISA.

Specifications

Antigen KLH
Applications ELISA
Classification Polyclonal
Conjugate DyLight 594
Formulation 50 mM Sodium Borate
Gene Alias keyhole limpet hemocyanin (KLH)
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Keyhole Limpet Hemocyanin.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.