Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLH Monoclonal Antibody (3B1), Rockland™

Mouse Monoclonal Antibody
$174.73 - $619.37
Specifications
Antigen | KLH |
---|---|
Clone | 3B1 |
Concentration | 1 mg/mL |
Applications | ELISA, Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0598
![]() ![]() |
Rockland Immunochemicals
200301E66 |
100 μg |
Each for $619.37
|
|
|||||
50-199-0599
![]() ![]() |
Rockland Immunochemicals
200301E66S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. KLH antibody is specific for KLH. Does not cross react with mouse heart or skin tissue sections. Cross-reactivity from other sources has not been determined.Specifications
KLH | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Anti-KLH (MOUSE) Monoclonal Antibody was produced in mouse by repeated immunizations with KLH (Keyhole Limpet Hemocyanin) followed by hybridoma development. | |
Primary | |
Antibody |
3B1 | |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Mouse | |
0.1M tris HCl with 0.5M NaCl and 0.1% sodium azide; pH 8 | |
IgM | |
-20°C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title