Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHDC3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP185198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
25ul
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP185198 0.1 mL
NB432331 25ul
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP185198 Supplier Novus Biologicals Supplier No. NBP185198
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLHDC3 Polyclonal specifically detects KLHDC3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen KLHDC3
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:10-1:20
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias hPeas, kelch domain containing 3, kelch domain-containing protein 3, PEASdJ20C7.3, Protein Peas, RP1-20C7.3, Testis intracellular mediator protein
Gene Symbols KLHDC3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:FHSATMLGSHMYVFGGRADRFGPFHSNNEIYCNRIRVFDTRTEAWLDCPPTPVLPEGRRSHSAFGYNGELYIFGGYNARLNRHFHDLWKFNPVSF
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 116138
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.