Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHDC3 Mouse anti-Mouse, Clone: A8-E8, Novus Biologicals™
SDP

Catalog No. NBP266919 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266919 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266919 Supplier Novus Biologicals Supplier No. NBP266919

Mouse Monoclonal Antibody

KLHDC3 Monoclonal specifically detects KLHDC3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen KLHDC3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A8-E8
Concentration 2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias hPeas, kelch domain containing 3, kelch domain-containing protein 3, PEASdJ20C7.3, Protein Peas, RP1-20C7.3, Testis intracellular mediator protein
Gene Symbols KLHDC3
Host Species Mouse
Immunogen Recombinant protein within Human KLHDC3 aa 121-382/ 382. (SwissProt: Q9BQ90 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 116138
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.