Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLHDC8A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | KLHDC8A |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15742220
![]() |
Novus Biologicals
NBP15742220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157422
![]() |
Novus Biologicals
NBP157422 |
100 μL |
Each for $487.50
|
|
|||||
Description
KLHDC8A Polyclonal specifically detects KLHDC8A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
KLHDC8A | |
Unconjugated | |
Rabbit | |
Q8IYD2 | |
55220 | |
Synthetic peptides corresponding to KLHDC8A (kelch domain containing 8A) The peptide sequence was selected from the C terminal of KLHDC8A. Peptide sequence PGKNKWEILPAMPTPRCACSSIVVKNCLLAVGGVNQGLSDAVEALCVSDS. | |
Primary |
Polyclonal | |
Purified | |
RUO | |
FLJ10748, kelch domain containing 8A, kelch domain-containing protein 8A | |
KLHDC8A | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title