Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL12/C3IP1 Antibody - BSA Free
SDP

Catalog No. NB12014233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB12014233 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB12014233 Supplier Novus Biologicals Supplier No. NB12014233

Chicken Polyclonal Antibody has been used in 2 publications

KLHL12/C3IP1 Polyclonal specifically detects KLHL12/C3IP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen KLHL12/C3IP1
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q53G59, Q8BZM0, Q8R2H4
Gene Alias C3IP1kelch-like protein 12, CUL3-interacting protein 1, DKIR, FLJ27152, kelch-like 12 (Drosophila), kelch-like protein C3IP1
Gene Symbols KLHL12
Host Species Chicken
Immunogen Synthetic peptide: KHAKKEREES LPNLLQYVRM PLLTPRYITD VIDAEPFIRC SLQCRDLVDE AKKFHLRPEL RSQMQGPRTR ARLGANEVLL VVGGFGSQQS PIDVVEKYDP KTQEWSFLPS ITRKRRYVAS VSLHDRIYVI GGYDGRSRLS SVECLDYTAD EDGVWYSVAP MNVRRGLAGA TTLGDMIYVS GGFDGSRRHT SMERYDPNID, corresponding to amino acids 205-404 of Human C3IP1.
Purification Method Affinity Purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 59349
Test Specificity C3IP1
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.