Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL25 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB356438 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356438 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356438 Supplier Novus Biologicals Supplier No. NBP177147MFV450

Chicken Polyclonal Antibody

KLHL25 Polyclonal antibody specifically detects KLHL25 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen KLHL25
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ectodermal-neural cortex 2, ENC-2BTB/POZ KELCH domain protein, ENC2ectoderm-neural cortex protein 2, FLJ12587, FLJ45015, kelch-like 25 (Drosophila), Kelch-like protein 25
Host Species Chicken
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the center of human ENC-2. The immunogen is located within amino acids 230 - 280 of ENC-2. Amino Acid Squence: SSEALMADERTKL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 64410
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.