Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL8 Antibody, Novus Biologicals™
SDP

Catalog No. NBP18034720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP18034720 20 μL
NBP180347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP18034720 Supplier Novus Biologicals Supplier No. NBP18034720UL

Rabbit Polyclonal Antibody

KLHL8 Polyclonal specifically detects KLHL8 in Human samples. It is validated for Western Blot.

Specifications

Antigen KLHL8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_065854
Gene Alias kelch-like 8 (Drosophila), kelch-like protein 8, KIAA1378FLJ46304
Gene Symbols KLHL8
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human KLHL8. Peptide sequence TVEAFDPVLNRWELVGSVSHCRAGAGVAVCSCLTSQIRDVGHGSNNVVDC.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57563
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.