Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL9 Antibody, Novus Biologicals™
SDP

Catalog No. NB395282 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB395282 25 μL
NBP254727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB395282 Supplier Novus Biologicals Supplier No. NBP25472725UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KLHL9 Polyclonal specifically detects KLHL9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen KLHL9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias FLJ13568, FLJ21815, kelch-like 9 (Drosophila), kelch-like protein 9, KIAA1354
Gene Symbols KLHL9
Host Species Rabbit
Immunogen This antibody was developed against a Recombinant Protein corresponding to amino acids:RYQHGIAVIGNFLYVVGGQSNYDTKGKTAVDTVFRFDPRYNKWMQVASLNE
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55958
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.