Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLHL9 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$480.74

Specifications

Antigen KLHL9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP155091
SDP
View Documents
Novus Biologicals
NBP155091
100 μL
Each for $480.74
Only null left
Add to Cart
 
NBP1550920
SDP
View Documents
Novus Biologicals
NBP15509120UL
20 μL N/A N/A N/A
Description

Description

KLHL9 Polyclonal specifically detects KLHL9 in Human samples. It is validated for Western Blot.
Specifications

Specifications

KLHL9
Polyclonal
Rabbit
Q9P2J3
55958
Synthetic peptides corresponding to KLHL9(kelch-like 9 (Drosophila)) The peptide sequence was selected from the middle region of KLHL9. Peptide sequence SALKGHLYAVGGRSAAGELATVECYNPRMNEWSYVAKMSEPHYGHAGTVY.
Primary
Western Blot
Unconjugated
RUO
FLJ13568, FLJ21815, kelch-like 9 (Drosophila), kelch-like protein 9, KIAA1354
KLHL9
IgG
69 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.