Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Klkbl4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | Klkbl4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15761720
![]() |
Novus Biologicals
NBP15761720UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157617
![]() |
Novus Biologicals
NBP157617 |
100 μL |
Each for $487.50
|
|
|||||
Description
Klkbl4 Polyclonal specifically detects Klkbl4 in Human samples. It is validated for Western Blot.Specifications
Klkbl4 | |
Polyclonal | |
Rabbit | |
Q6PEW0 | |
221191 | |
Synthetic peptides corresponding to KLKBL4 The peptide sequence was selected from the C terminal of KLKBL4 (NP_001073961). Peptide sequence EASVQPLYYDYYGGEVGEGRIFAGQNRLYQPEEIILVSFVLVFFCSSI. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
Cancer/testis antigen 67, CT67Plasma kallikrein-like protein 4, FLJ25339, KLKBL4inactive serine protease 54, protease, serine, 54 | |
PRSS54 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title